Order Entry
Switzerland
ContactUsLinkComponent
Anti-COX4 + COX4L2 Mouse Monoclonal Antibody [clone: 10G8D12C12] (Alexa Fluor® 488)
Anti-COX4 + COX4L2 Mouse Monoclonal Antibody [clone: 10G8D12C12] (Alexa Fluor® 488)
Codice catalogo ABCAAB198590-100
Nome del fornitore:  Abcam
undefined
Anti-COX4 + COX4L2 Mouse Monoclonal Antibody [clone: 10G8D12C12] (Alexa Fluor® 488)
Codice catalogo ABCAAB198590-100
Nome del fornitore:  Abcam

Specifiche

  • Taglia:
    100 µl
  • Tipo di conf.:
    Vial

Specifiche

Informazioni su questo articolo

Alexa Fluor® 488 Mouse monoclonal [10G8D12C12] to COX4 + COX4L2.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: COX4 + COX4L2
Clonality: Monoclonal
Clone: 10G8D12C12
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human