Anti-Thymidine Phosphorylase Mouse Monoclonal Antibody [clone: P-GF.44C]
Codice catalogo ABCAAB218381-100
Nome del fornitore: Abcam
Specifiche
- Seq:1286
- Taglia:100 µg
- Tipo di conf.:Vial
Specifiche
Informazioni su questo articolo
Mouse monoclonal [P-GF.44C] to Thymidine Phosphorylase.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Thymidine Phosphorylase
Clonality: Monoclonal
Clone: P-GF.44C
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human