Anti-Loricrin Rabbit Monoclonal Antibody [clone: EPR7149(2)]
: Abcam
New Product
Rabbit monoclonal [EPR7149(2)] to Loricrin - C-terminal.
Type: Primary
Antigen: Loricrin
Clonality: Monoclonal
Clone: EPR7149(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat
Frequently Bought Together