Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD6/T12 Mouse Monoclonal Antibody [clone: C6/372 + 3F7B5]
Anti-CD6/T12 Mouse Monoclonal Antibody [clone: C6/372 + 3F7B5]
Codice catalogo ABCAAB199008-500
Nome del fornitore:  Abcam
undefined
Anti-CD6/T12 Mouse Monoclonal Antibody [clone: C6/372 + 3F7B5]
Codice catalogo ABCAAB199008-500
Nome del fornitore:  Abcam

Specifiche

  • Seq:
    1418
  • Taglia:
    500 µl
  • Tipo di conf.:
    Vial

Specifiche

Informazioni su questo articolo

Mouse monoclonal [C6/372 + 3F7B5] to CD6/T12.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD6/T12
Clonality: Monoclonal
Clone: C6/372 + 3F7B5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human