Order Entry
Switzerland
ContactUsLinkComponent
Anti-MelanA Mouse Monoclonal Antibody [clone: A103-M27C10-M29E3]
Anti-MelanA Mouse Monoclonal Antibody [clone: A103-M27C10-M29E3]
Catalog # ABCAAB200544-500
Supplier:  Abcam
undefined
Anti-MelanA Mouse Monoclonal Antibody [clone: A103-M27C10-M29E3]
Catalog # ABCAAB200544-500
Supplier:  Abcam

Specifications

  • Seq:
    1418
  • Taglia:
    500 µl
  • Tipo di conf.:
    Vial

Specifications

About this item

Mouse monoclonal [A103-M27C10-M29E3] to MelanA.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: MelanA
Clonality: Monoclonal
Clone: A103-M27C10-M29E3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human