Anti-ATP synthase C Rabbit Monoclonal Antibody [clone: EPR13907] (Alexa Fluor® 488)
Codice catalogo ABCAAB210456-100
Nome del fornitore: Abcam
Specifiche
- Taglia:100 µl
- Tipo di conf.:Vial
Specifiche
Informazioni su questo articolo
Alexa Fluor® 488 Rabbit monoclonal [EPR13907] to ATP synthase C.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ATP synthase C
Clonality: Monoclonal
Clone: EPR13907
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human