Weighing scoop
155 mm, 9 mm
Codice catalogo USBE3700
Nome del fornitore: USBECK Laborgeräte
Vai a
Product Details & Documents
Nickel plated brass, with plastic handle.
Specifiche
- Larghezza:9 mm
- Lunghezza:155 mm
Nickel plated brass, with plastic handle.