Orders LinkContactUsLinkComponent
Molecular models of elements, Molymod®
Marketsource
Molecular models of elements, Molymod®
Marketsource
Molecular models of elements, Molymod®
Molecular models of elements, Molymod®
Molymod is the original, unique, dual-scale system of high quality molecular models.
Product Family Options
  • Descrizione
    Colore
    N. di fori
    Ø
  • Metal
    grigio
    1
    20 mm
    1 * 10 ST
    1 * 10 ST
    Marketsource
    Marketsource
  • Halogen
    verde
    1
    17 mm
    1 * 10 ST
    1 * 10 ST - SFr. 2.80
    SFr. 2.80
    1 * 10 ST
       SFr. 2.80
    Marketsource
    Marketsource
  • Oxygen
    rosso
    2
    23 mm
    1 * 10 ST
    1 * 10 ST
    Marketsource
    Marketsource
  • Sulphur
    giallo
    2
    23 mm
    1 * 10 ST
    1 * 10 ST
    Marketsource
    Marketsource
  • Metal
    grigio
    2
    23 mm
    1 * 10 ST
    1 * 10 ST
    Marketsource
    Marketsource
  • Nitrogen
    blu
    3
    23 mm
    1 * 10 ST
    1 * 10 ST
    Marketsource
    Marketsource
  • Phosphorus
    porpora
    5
    23 mm
    1 * 10 ST
    1 * 10 ST
    Marketsource
    Marketsource
  • Nitrogen
    blu
    5
    23 mm
    1 * 10 ST
    1 * 10 ST
    Marketsource
    Marketsource
  • Sulphur
    giallo
    6
    23 mm
    1 * 10 ST
    1 * 10 ST
    Marketsource
    Marketsource
  • Hydrogen
    bianco
    1
    17 mm
    1 * 10 ST
    1 * 10 ST
    Marketsource
    Marketsource
  • Bromine
    arancione
    1
    17 mm
    1 * 10 ST
    1 * 10 ST
    Marketsource
    Marketsource
  • Carbon
    nero
    4
    23 mm
    1 * 10 ST
    1 * 10 ST
    Marketsource
    Marketsource
  • Halogen
    verde
    1
    20 mm
    1 * 10 ST
    1 * 10 ST
    Marketsource
    Marketsource
  • Nitrogen
    blu
    4
    23 mm
    1 * 10 ST
    1 * 10 ST
    Marketsource
    Marketsource
  • Oxygen
    rosso
    4
    23 mm
    1 * 10 ST
    1 * 10 ST
    Marketsource
    Marketsource
  • Sulphur
    giallo
    4
    23 mm
    1 * 10 ST
    1 * 10 ST
    Marketsource
    Marketsource
  • Phosphorus
    porpora
    4
    23 mm
    1 * 10 ST
    1 * 10 ST
    Marketsource
    Marketsource
  • Silicon
    grigio
    4
    23 mm
    1 * 10 ST
    1 * 10 ST
    Marketsource
    Marketsource
  • Carbon
    nero
    3
    23 mm
    1 * 10 ST
    1 * 10 ST
    Marketsource
    Marketsource
  • Nitrogen
    blu
    2
    23 mm
    1 * 10 ST
    1 * 10 ST