Safety spectacles, Prism
Anti-UV, anti-scratch, anti-fog, trasparente, Clear PC
Codice catalogo BOLEPRIPSI
Nome del fornitore: Bollé Safety
Vai a
Product Details & Documents
The model Prism with dual lens offers a high level of protection and its frame is entirely composed of ultra resistant PC.
- Panoramic vision
- Sporty design and provides upper protection
- Comfortable with straight temples
Weight: 23 g
Certifications: EN 166; EN 170
Informazioni per l'ordine: Supplied with adjustable cord.
Specifiche
- Tipo:Anti-UV, anti-scratch, anti-fog
- Colore:trasparente
- Materiale lente:Clear PC