Face shield, sphere
Anti-UV (2C-1.2), short circuit electric arcs protection, PC, 205×394 mm
Codice catalogo 111-1024
Nome del fornitore: Bollé Safety
Vai a
Product Details & Documents
This protective face shield, with PC lens, provides protection against UV rays and short circuit electric arcs.
- 180° field of vision
- Easy to fit due to height- and width-adjustable headgear and headband
- Face shield adapts to all head shapes for all uses
Weight: 355 g
Certifications: EN 166.
Specifiche
- Tipo:Anti-UV (2C-1.2), short circuit electric arcs protection
- L×L:205×394 mm
- Materiale:PC
Accessories
Informazioni sul prodotto
- Codice catalogoTipoMaterialeDisponibilitàPrice
- Replacement visor lens, 205×394 mmReplacement visor lens, 205×394 mm111-1003Anti-UV, short circuit electric arcs protectionPCLog in per disponibilità1 * 1 ST - SFr. 19.00