Capillary columns, OPTIMA® 210
Supplier: MACHEREY-NAGEL
Certificates
About this item
Medium polar GC phase.
- Recommended for environmental analyses, especially for o-, m- and p-substituted aromatic hydrocarbons
- Maximum temperature for isothermal operation 260 °C
- Maximum temperature for short isotherms in a temperature program 280 °C
50% trifluoropropyl – 50% methylpolysiloxane phase.
USP code: G6.
Frequently Bought Together