Adapters, socket /screwthread
29/32 NS, SVL 30
Codice catalogo BIBB670-05
Nome del fornitore: SciLabware
Vai a
Product Details & Documents
Adapters, borosilicate glass 3.3. Facilitates connection between SVL® and Quickfit® glassware. Features an SVL® screwthread and Quickfit® ground glass socket. Thread accepts SVL® screwcaps and couplings. Socket fits Quickfit® cones.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Specifiche
- Cono femmina:29/32 NS
- Filettatura:SVL 30