Refrigerators, wine storage cabinets
Nome del fornitore: Liebherr
Certificati
Informazioni su questo articolo
Cabinets designed for storage and ageing of wines with a protected environment.
- Digital temperature display
- Height-adjustable wooden grid shelves
- High-quality SwingLine design, reversible doors and subsequent lock
- Forced air cooling and fresh air supply via activated charcoal filter
- Vibration-free compressors
Frequently Bought Together