Analogue stopwatch, bench top
HECH41979010
- Colore:bianco
- Descrizione:Table/wall stopclock
- Conf:1
Electronic stopclock for bench top use or for wall mounting.
Timing capacity: 0 to 60 seconds, 0 to 60 minutes (scale outside), 0 to 100 / 100 minutes (scale inside).
: Including battery (AA) 1,5 V.