ContactUsLinkComponent
Anion exchange resins, Toyopearl® and TSK-GEL®
Anion exchange resins, Toyopearl® and TSK-GEL®
Supplier:  TOSOH Bioscience
Certificates
undefined
Anion exchange resins, Toyopearl® and TSK-GEL®
Supplier:  TOSOH Bioscience
Certificates
Specifications
  • Quantità di gruppi ionici:
    200 - 300 µeq/ml
  • Granulometria media:
    20 - 50 µm
  • Resistenza alla pressione:
    3 bar
  • Tipo:
    SuperQ-650S
  • pH di esercizio:
    2 - 13
Specifications
Related Information
Frequently Bought Together