Orders LinkContactUsLinkComponent
Pasteur pipettes, disposable
Pasteur pipettes, disposable
Pasteur pipettes, disposable
Certificates
Pasteur pipettes, disposable
Pasteur pipettes, disposable
4 Options Available Below
Product Details & Documents
These disposable Pasteur pipettes are made of soda glass.
  • Long tapered tip
  • With constriction for cotton plug
  • Available with or without cotton plug
Certifications: ISO 7712
Documents
Product Family Options
Short form
Product Information
  • Descrizione
    Capacità
    Ø est
    Lunghezza
    Confezione
    Availability
    Price
  • Pre-plugged
    2 ml
    7 mm
    150 mm
    250/box, 4 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - SFr. 146.00
    SFr. 146.00
    1 * 1.000 ST
       SFr. 146.00
  • Non plugged
    2 ml
    7 mm
    150 mm
    250/box
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - SFr. 77.00
    SFr. 77.00
    1 * 1.000 ST
       SFr. 77.00
Long form
Product Information
  • Descrizione
    Capacità
    Ø est
    Lunghezza
    Confezione
    Availability
    Price
  • Pre-plugged
    2 ml
    7 mm
    230 mm
    250/box
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - SFr. 37.70
    SFr. 37.70
    1 * 250 ST
       SFr. 37.70
  • Non plugged
    2 ml
    7 mm
    230 mm
    250/box, 4 boxes/case
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - SFr. 82.00
    SFr. 82.00
    1 * 1.000 ST
       SFr. 82.00