L(+)-Lysine dihydrochloride
Catalog # CAMBULM-8766-0.1
Supplier: CAMBRIDGE ISOTOPE LABS
CAS Number:
Specifications
- Tipo di conf.:Bottiglia di vetro
- Conf:0,1 g
- Seq:3020
Specifications
About this item
:
:219.11 g/mol
Melting Pt:200.2 °C
EINECS:211-518-3
CAS Number:657-26-1
Formula:C₆H₁₆Cl₂N₂O₂
MDL Number:MFCD00066389
Melting Pt:200,2 °C