Solid phase extraction cartridges, Bond Elut™ Certify II
Supplier: VARIAN
Certificates
About this item
Bond Elut™ Certify II was developed specifically for the rapid and effective extraction of acidic drugs and metabolites from urine and other biological matrices. Certify II is a mixed-mode cartridge packed with non-polar C8 and strong anion exchange (SAX) sorbent. It has been optimised for acidic drugs such as 11-nor-delta-9-tetrahydrocannibinol-carboxylic acid, salicylic acid, ibuprofen, acetaminophen and other compounds that possess both non-polar and anionic characteristics.
- Ideal for non-polar and anionic compounds
- Optimised for acidic drug analysis
- Bimodal, non-polar and strong anion exchange
Specifications
- Catalog No:
- 551-0421
- 551-0423
- VARI12102088
- 551-0420
- 551-0418
- VARI12105031
- VARI12113063
- VARI12102084
- VARI12102080
- VARI102818C
- VARI12113051
- Carico di carbonio:
- 8,6%
- 8,6%
- 8,6%
- 8,6%
- 8,6%
- 8,6%
- 8,6%
- 8,6%
- 8,6%
- 8,6%
- 8,6%
- Con endcapping:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Area superficiale:
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g