Cut resistant sleeves, HyFlex® 72-265
ANSE72-265
: Ansell
- Applicazioni per abiti:Industrial
- Risk area:Cuts
- Colore:grigio
- Lunghezza:380 mm
- Seq:05
- Tessuto:Fiber glass/polyester/Dyneema®
- Conf:1
These sleeves contain a very high proportion of Dyneema® fibres which are as strong as steel fibres but much lighter and more flexible.
- Sleeves provide optimal protection from cuts and abrasions
- Light weight and comfortable fit ensure high levels of user acceptance
Ideal for use in plastics injection and moulding, paper processing and metal fabrication.
: EN 388:2016 (3X4XC), Cat. II