Orders LinkContactUsLinkComponent
Ductless ventilation systems, Protector®XVS™
Ductless ventilation systems, Protector®XVS™
Supplier:   LABCONCO
Ductless ventilation systems, Protector®XVS™
Supplier:  LABCONCO
Certificates
Ductless ventilation systems, Protector®XVS™
4 Options Available Below
Product Details & Documents
Ductless ventilation systems with glass tops and sides and low profiles are well suited for educational settings. Clean-Sweep air foil and zone-perforated rear baffle maximise containment.
These ventilation stations can be used as light duty fume hoods, science classroom work stations, solvent cleaning work bays, weighing stations or for pathology and cytology.
Protector XVS ventilation stations can be ducted to the outside or connected to the FilterMate portable exhauster for HEPA and/or carbon filtered exhaust back into the laboratory.
Certifications: Conforms to ANSI Z9.5, Modified ASHRAE 110, SEFA 1.
Specifications for Product Family
Specifications
  • Catalog No:
  • LBCP4863010
  • LBCP4863030
  • LBCP4864010
  • LBCP4864030
  • Luminosità ( lux ):
  • non incluso
Specifications
Product Family Options
Product Information
  • Descrizione
    Esterno L×P×A
    Peso
    Availability
    Price
  • Cappa chimica, Protector® XVS™, da banco, (larghezza nominale 76,2 mm)
    914×737×578 mm
    31,8 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    LBCP4863010
    Luminosità ( lux )
    non incluso
  • 914×736×812 mm
    36,3 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    LBCP4863030
    Luminosità ( lux )
  • 1219×736×577 mm
    40,8 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    LBCP4864010
    Luminosità ( lux )
  • 1219×736×812 mm
    45,3 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    More
    Specifications
    Cat. No.
    LBCP4864030
    Luminosità ( lux )
Accessories
Product Information
  • Catalog #
    Descrizione
    L×P×A
    Availability
    Price
  • LBCP3908401
    Solid epoxy work surface, black, 910 mm width, 33 kg
    910×680×25 mm
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Descrizione
    L×P×A
    Availability
    Price
  • LBCP3908400
    Solid epoxy work surface, black, 610 mm width, 22 kg
    610×680×25 mm
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Descrizione
    L×P×A
    Availability
    Price
  • LBCP3908402
    Solid epoxy work surface, black, 1220 mm width, 44 kg
    1220×680×25 mm
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Descrizione
    L×P×A
    Availability
    Price
  • LBCP3925000
    Utility shelves attachs to the rear baffle perforations, has a scalloped front edge to support scalpels, forceps and other supplies, 2,3 kg
    — mm
    Log in for availability
    1 * 1 ST - SFr. 217.00
Product Information
  • Catalog #
    Descrizione
    L×P×A
    Availability
    Price
  • LBCP3908801
    Digital airflow monitorwith epoxy coated steel housing, Gaurdian 1000, 4,5 kg
    — mm
    Log in for availability
    1 * 1 ST - SFr. 3’770.00
Product Information
  • Catalog #
    Descrizione
    L×P×A
    Availability
    Price
  • LBCP3746712
    Telescoping base stands with castors, nominal width of 101,6 mm, 1219×1016 mm, 38 kg
    — mm
    Log in for availability
    1 * 1 ST - SFr. 2’240.00
Product Information
  • Catalog #
    Descrizione
    L×P×A
    Availability
    Price
  • LBCP3912401
    Upper exhaust transition adaptor, 152,4 mm width, 2,3 kg
    — mm
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Descrizione
    L×P×A
    Availability
    Price
  • 135-1092
    Portable exhauster, FilterMate™, with available HEPA filter, 22,7 kg
    360×580×580 mm
    Log in for availability
    1 * 1 ST - SFr. 5’760.00
Product Information
  • Catalog #
    Descrizione
    L×P×A
    Availability
    Price
  • 135-1080
    Portable exhauster, FilterMate™, with available ammonia-amine, formaldehyde, organic vapor filter, 22,7 kg
    360×580×580 mm
    Log in for availability
    1 * 1 ST - SFr. 4’960.00
Product Information
  • Catalog #
    Descrizione
    L×P×A
    Availability
    Price
  • 135-1081
    Portable exhauster, FilterMate™, with HEPA filter included, 22,7 kg
    470×580×630 mm
    Log in for availability
    1 * 1 ST - SFr. 6’000.00
Product Information
  • Catalog #
    Descrizione
    L×P×A
    Availability
    Price
  • 135-1093
    Portable exhauster, FilterMate™, with available ammonia-amine, formaldehyde, organic vapor and HEPA filter, 22,7 kg
    380×580×580 mm
    Log in for availability
    1 * 1 ST - SFr. 6’260.00
Product Information
  • Catalog #
    Descrizione
    L×P×A
    Availability
    Price
  • 135-1082
    Portable exhauster, FilterMate™, with available ammonia-amine, formaldehyde, organic vapor filter, 41 kg
    380×580×580 mm
    Log in for availability
    1 * 1 ST - SFr. 5’390.00
Product Information
  • Catalog #
    Descrizione
    L×P×A
    Availability
    Price
  • 135-1091
    Organic carbon filter, 7,7 kg
    180×180×210 mm
    Log in for availability
    1 * 1 ST - SFr. 521.00
Product Information
  • Catalog #
    Descrizione
    L×P×A
    Availability
    Price
  • 135-1078
    Formaldehyde carbon filter, 9,5 kg
    457×457×530 mm
    Log in for availability
    1 * 1 ST - SFr. 895.00
Product Information
  • Catalog #
    Descrizione
    L×P×A
    Availability
    Price
  • 135-1079
    Ammonia carbon filter, 9,5 kg
    457×457×530 mm
    Log in for availability
    1 * 1 ST - SFr. 895.00
Product Information
  • Catalog #
    Descrizione
    L×P×A
    Availability
    Price
  • 135-1077
    Replacement HEPA filter, 99,99 % on particles 0,3 micron in size
    457×457×83,82 mm
    Log in for availability
    1 * 1 ST - SFr. 895.00
Product Information
  • Catalog #
    Descrizione
    L×P×A
    Availability
    Price
  • LBCP3776002
    Replacement bag for bag-in/ bag out HEPA filter
    — mm
    Log in for availability
    1 * 1 ST - SFr. 504.00
Product Information
  • Catalog #
    Descrizione
    L×P×A
    Availability
    Price
  • LBCP6925700
    Ammonia detector tube, 0,5 kg
    — mm
    Log in for availability
    1 * 1 ST - SFr. 161.00
Product Information
  • Catalog #
    Descrizione
    L×P×A
    Availability
    Price
  • LBCP6926000
    Formaldehyde detector tube, 0,5 kg
    — mm
    Log in for availability
    1 * 1 ST - SFr. 197.00
Product Information
  • Catalog #
    Descrizione
    L×P×A
    Availability
    Price
  • LBCP3908407
    Solid epoxy work surface, black, 1830 mm width, 63 kg
    1830×680×25 mm
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Descrizione
    L×P×A
    Availability
    Price
  • LBCP3908410
    Solid epoxy work surface, black, 1830 mm width, 84 kg
    1830×900×25 mm
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Descrizione
    L×P×A
    Availability
    Price
  • LBCP3908800
    Guardian™ 115 V, 60 Hz
    1000 digital airflow monitor for protector XVS and demonstration hood
    — mm
    Log in for availability
    1 * 1 ST - SFr. 3’770.00
Product Information
  • Catalog #
    Descrizione
    L×P×A
    Availability
    Price
  • LBCP9743209
    Guardian™ 230 V, 50/60 Hz digital airflow monitor for basic™ hoods
    — mm
    Log in for availability
    1 * 1 KIT
Product Information
  • Catalog #
    Descrizione
    L×P×A
    Availability
    Price
  • LBCP9743201
    Guardian™ 115 V, 50/60 Hz digital airflow monitor for fiberglass 30 hoods
    — mm
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • LBCP3811102
    Accessory shelf, 4'
    Log in for availability
    1 * 1 ST - SFr. 369.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • LBCP3811103
    Accessory shelf, 5'
    Log in for availability
    1 * 1 ST - SFr. 462.00
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..