Cell culture flasks with reclosable lid
: TPP
Polystyrene tissue culture flasks with multi-use reclosable liquid-tight lid.
- Surface activated, uniform flat growth surface
- Crystal clear transparency
- Geometry of the neck reduces risk of media slop over
- Complete access to flask chamber from above
- Stable and slip-free stacking of multiple flasks
Flasks are available only with filter screw caps. If a vented screw cap is used, a build-up of more than 0,03 bar inside the flask could press open the lid.
: ISO 9001:2008 certified.
: Optional barrier which creates a growth area of 115 cm² can be ordered separately.
Frequently Bought Together