Solid phase extraction cartridges, Bond Elut™ C8
About this item
Bond Elut™ C8 is very similar in properties to C18, but is not as retentive for non-polar compounds, due to its shorter hydrocarbon chain, and therefore reduced carbon loading. C8 is an excellent replacement for C18 when analytes are too strongly retained for effective elution. The potential for polar interactions is somewhat higher than for C18 because there is less coverage of the silica surface. These polar interactions are not, however, a significant property of C8.
- Excellent for strongly-retained analytes
- Polar interactions not significant
- Less retentive than C18
23 Options Available Below
Product Details & Documents
Bond Elut™ C8 is very similar in properties to C18, but is not as retentive for non-polar compounds, due to its shorter hydrocarbon chain, and therefore reduced carbon loading. C8 is an excellent replacement for C18 when analytes are too strongly retained for effective elution. The potential for polar interactions is somewhat higher than for C18 because there is less coverage of the silica surface. These polar interactions are not, however, a significant property of C8.
- Excellent for strongly-retained analytes
- Polar interactions not significant
- Less retentive than C18
Specifications for Product Family
Specifications
- Catalog No:
- VARI12113002
- VARI12113075
- VARI12113025
- VARI12113028
- 549-0788
- 549-0807
- 551-0301
- 549-0872
- 549-0798
- 551-0313
- 549-0873
- 551-0316
- 549-0786
- 549-0820
- 549-0825
- 549-0827
- 551-0374
- 551-0381
- 551-0428
- VARI75403050
- 551-0427
- VARI7540302C
- VARI7550301C
- Carico di carbonio:
- 12,2%
- 12,2%
- 12,2%
- 12,2%
- 12,2%
- 12,2%
- 12,2%
- 12,2%
- 12,2%
- 12,2%
- 12,2%
- 12,2%
- 12,2%
- 12,2%
- 12,2%
- 12,2%
- 12,2%
- 12,2%
- 12,2%
- 12,2%
- 12,2%
- 12,2%
- 12,2%
- Con endcapping:
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Area superficiale:
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
Specifications
Product Family Options
LRC cartridges
Product Information
- DescrizioneFaseDimensioni poriDimensioni particelleCapacitàPeso lettoAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.VARI12113002Carico di carbonio12,2%Con endcappingSìArea superficiale500 m²/g - Specifications:
More
Specifications
Cat. No.VARI12113075Carico di carbonio12,2%Con endcappingSìArea superficiale500 m²/g - Specifications:
More
Specifications
Cat. No.VARI12113025Carico di carbonio12,2%Con endcappingSìArea superficiale500 m²/g - Specifications:
More
Specifications
Cat. No.VARI12113028Carico di carbonio12,2%Con endcappingSìArea superficiale500 m²/g
Straight barrel cartridges
Product Information
- DescrizioneFaseDimensioni poriDimensioni particelleCapacitàPeso lettoAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.549-0788Carico di carbonio12,2%Con endcappingSìArea superficiale500 m²/g - Specifications:
More
Specifications
Cat. No.549-0807Carico di carbonio12,2%Con endcappingSìArea superficiale500 m²/g - Specifications:
More
Specifications
Cat. No.551-0301Carico di carbonio12,2%Con endcappingSìArea superficiale500 m²/g - Specifications:
More
Specifications
Cat. No.549-0872Carico di carbonio12,2%Con endcappingSìArea superficiale500 m²/g - Specifications:
More
Specifications
Cat. No.549-0798Carico di carbonio12,2%Con endcappingSìArea superficiale500 m²/g - Specifications:
More
Specifications
Cat. No.551-0313Carico di carbonio12,2%Con endcappingSìArea superficiale500 m²/g - Specifications:
More
Specifications
Cat. No.549-0873Carico di carbonio12,2%Con endcappingSìArea superficiale500 m²/g - Specifications:
More
Specifications
Cat. No.551-0316Carico di carbonio12,2%Con endcappingSìArea superficiale500 m²/g - Specifications:
More
Specifications
Cat. No.549-0786Carico di carbonio12,2%Con endcappingSìArea superficiale500 m²/g - Specifications:
More
Specifications
Cat. No.549-0820Carico di carbonio12,2%Con endcappingSìArea superficiale500 m²/g - Specifications:
More
Specifications
Cat. No.549-0825Carico di carbonio12,2%Con endcappingSìArea superficiale500 m²/g - Specifications:
More
Specifications
Cat. No.549-0827Carico di carbonio12,2%Con endcappingSìArea superficiale500 m²/g
Bond Elut™ Jr
Product Information
- DescrizioneFaseDimensioni poriDimensioni particelleCapacitàPeso lettoAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.551-0374Carico di carbonio12,2%Con endcappingSìArea superficiale500 m²/g - Specifications:
More
Specifications
Cat. No.551-0381Carico di carbonio12,2%Con endcappingSìArea superficiale500 m²/g
VersaPlate™ formats
Product Information
- DescrizioneFaseDimensioni poriDimensioni particelleCapacitàPeso lettoAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.551-0428Carico di carbonio12,2%Con endcappingSìArea superficiale500 m²/g - Specifications:
More
Specifications
Cat. No.VARI75403050Carico di carbonio12,2%Con endcappingSìArea superficiale500 m²/g - Specifications:
More
Specifications
Cat. No.551-0427Carico di carbonio12,2%Con endcappingSìArea superficiale500 m²/g - Specifications:
More
Specifications
Cat. No.VARI7540302CCarico di carbonio12,2%Con endcappingSìArea superficiale500 m²/g - Specifications:
More
Specifications
Cat. No.VARI7550301CCarico di carbonio12,2%Con endcappingSìArea superficiale500 m²/g