Multi-position magnetic stirrers, MultiMagStir Genie®
About this item
The programmable revolutionary 4-position MultiMagStir Genie provides low to high speed stirring and editable memory profiles.
- Three power levels for various sample viscosities
- Manual mode provides stir, stir revere, and time functions
- Timer: 1 second to 99 hours
- Memory up to 4 user-defined profiles
- With selectable ramp rate of 5 to 300 min−1/sec
Microprocessor controls offer precision and reproducibility without internal heat build-up. The stirrer incorporates a feature to program user profiles which is ideal for repetitive protocols or for use by different users. Maximum load capacity is 10 kg.
Certifications: ETL listed.
0 Options Available Below
Product Details & Documents
The programmable revolutionary 4-position MultiMagStir Genie provides low to high speed stirring and editable memory profiles.
- Three power levels for various sample viscosities
- Manual mode provides stir, stir revere, and time functions
- Timer: 1 second to 99 hours
- Memory up to 4 user-defined profiles
- With selectable ramp rate of 5 to 300 min−1/sec
Microprocessor controls offer precision and reproducibility without internal heat build-up. The stirrer incorporates a feature to program user profiles which is ideal for repetitive protocols or for use by different users. Maximum load capacity is 10 kg.
Certifications: ETL listed.
Specifications for Product Family
Specifications
- Catalog No:
- SCIISI-3002
- SCIISI-3003
- Modello:
- MultiMagStir Genie®
- MultiMagStir Genie®
- Numero di posizioni di agitazione:
- 4
- 4
- Capacità di agitazione max. per agitatore H₂O:
- 1000 ml ×4 (or max 3×1 L spinner flasks)
- 1000 ml ×4 (or max 3×1 L spinner flasks)
- Intervallo di velocità:
- 10 - 1200 min⁻¹
- 10 - 1200 min⁻¹
- Materiale della piastra:
- ABS/PC
- ABS/PC
- L×P×A:
- 325×375×48 mm
- 325×375×48 mm
- Peso:
- 3,2 kg
- 3,2 kg