D(+)-α-Acetobromoglucose
Glass bottle for solids, 25 g
Catalog # MOLE27442186-25G
Marketsource
Supplier: Molekula
D(+)-α-Acetobromoglucose
Catalog # MOLE27442186-25G
Supplier: Molekula
Marketsource
CAS Number: 572-09-8
Jump to:
Product Details & Documents
Synonyms: 2,3,4,6-Tetra-O-acetyl-α-D-glucopyranosyl bromide, Bromo-2,3,4,6-Tetra-o-Acetyl-α-D-Glucopyranose, Acetobromo glucose, (2R,3R,4S,5R,6R)-2-(Acetoxymethyl)-6-bromotetrahydro-2H-pyran-3,4,5-triyl triacetate, α-Acetobromoglucose, Acetobromo-α-D-glucose
- CAS Number:572-09-8
- Formula:C₁₄H₁₉BrO₉
- MDL Number:MFCD00063254
- Molecular Weight:411.2 g/mol
- EINECS:209-339-0
Specifications
Specifications
- Tipo di conf.:Glass bottle for solids
- Conf:25 g



