Order Entry
Switzerland
ContactUsLinkComponent
1,1,1-Tricloroetano-D3 (98% D)
Irritant
1,1,1-Tricloroetano-D3 (98% D)
Catalog # PROODLM1974-1
CAS Number:  
Irritant
1,1,1-Tricloroetano-D3 (98% D)
Catalog # PROODLM1974-1
CAS Number:  

Specifications

  • Tipo di conf.:
    Bottiglia di vetro
  • Conf:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Storage Temperature:Ambiente
UN:2831
ADR:6.1,III
 :GHS07
CAS Number:2747-58-2
Formula:Cl₃CCD₃