ContactUsLinkComponent
Dimroth condensers for extractors
  HECH41154045
undefined
Dimroth condensers for extractors
  HECH41154045
  • Dimensione cono:
    45/40 NS
  • Per:
    Soxleth extractor 100 - 250 ml
  • Conf:
    1
DURAN® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Dimroth condensers for extractors, for condensing vapours. Water connection with two glass olives.