Woulff bottles
1000 ml, 110 mm, 185 mm, two side necks with GL 18 thread, center neck with GL 25 thread
Codice catalogo WITG5.802.001
Nome del fornitore: witeg Labortechnik
Vai a
Product Details & Documents
Made of borosilicate glass 3.3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion gives relatively high resistance to temperature changes
Two side necks with GL 18 thread, center neck with GL 25 thread. With bottom tubulature with screw cap and olive for tubing connection.
Specifiche
- Ø est:110 mm
- Capacità:1000 ml
- Altezza:185 mm
- Collo:two side necks with GL 18 thread, center neck with GL 25 thread