Reusable half masks, MX/PF
111-0548
- Tipo:MX/PF F 950
- Taglia:U*
- Conf:1
The half mask MX/PF is designed to work when the type of protection depends on the cartridges connected, gases, particles or combined. The APF is evaluated from 12 to 50.
- Easy assembly
- Silicone facepiece for additional comfort and easy decontamination
- Triple flange seal
- Wide, horizontal skin contact with pleated design
- Good fit accross a wide variety of facial geometries
The half mask SPERIAN MX PF can be used only with certified T SERIES cartridges (following the standard EN 141 , EN 143)
Recommended for long term use