Tree-pruning harness
1,78 kg
Codice catalogo DALL1002927
Nome del fornitore: HONEYWELL SAFETY
Vai a
Product Details & Documents
Commonly used in conjunction with a climbing line, spikes and a work-positioning lanyard, these 2-point full body harnesses are suitable for heavier tree-pruning operations, for example when using a chainsaw. They feature a positioning belt, seat strap, rear anchorage for fall arrest and a sternal anchorage for the climbing line.
- Climber is positioned against the trunk and is not required to move around the tree
- Shoulder straps evenly distribute the weight of the tools (e.g. chainsaws) through the harness
- Rigid seat strap provides a comfortable and safer position during work in suspension from the climbing line
- Wide, padded belt is used in conjunction with the positioning lanyard around the tree-trunk
Specifiche
- Peso:1,78 kg