Spectra/Por® Micro-dialysis Hollow Fibers
Codice catalogo SPEC132294
Nome del fornitore: Spectrum Laboratories
Specifiche
- Descrizione:Membrane in fibra cava per microdialisi, in vivo, in cellulosa
- Conf:20
Specifiche
Informazioni su questo articolo
The implantation of individual micro-dialysis hollow fibers is an ideal step in the recovery or introduction of micro-quantity biologicals or pharmaceutical materials in a localized tissue environment. Following sample injection into the lumen via syringe, the fiber ends may be sealed with epoxy for implantation.
Supplied in cut lengths of 152 mm (6").