Combs for Max Submarine Horizontal Electrophoresis Unit, He 99X
Catalog # HOEFHE91-BK
Supplier: HOEFER ELECTROPHORESIS
Specifications
- Descrizione:Comb back and 2 screws
- Seq:01
- Conf:1
Specifications
About this item
All combs require HE-91-BK comb back, ordered separately.