Wire, Regular Erecta Self®
Wire shelf, Erecta Shelf®, with adjustable feet (has to be assembled), load capacity 363 kg, Cromata, 3,7 kg
Catalog # METO1242C
Supplier: Metro International
Jump to:
Product Details & Documents
Chrome plated wire shelves is an easy-to-assemble system that puts space to work practically anywhere.
Specifications
- Descrizione:Wire shelf, Erecta Shelf®, with adjustable feet (has to be assembled), load capacity 363 kg
- Materiale:Cromata
- Peso:3,7 kg