Multi-channel pipettes, mechanical, variable volume, WITOPET-Multi
These multi-channel pipettes are designed for comfort over long term repetitive pipetting. 8 or 12 identical volumes for titration plates.
- Highest possible accuracy and precision
- Easy readable digital volume display
- Volume ranges colour coded on the push button
- Volumes steplessly adjustable
- Tip ejector can be blocked to avoid mis-application when using the instrument over large series
: Each pipette is individually tested and delivered with a test certificate.
These multi-channel pipettes are designed for comfort over long term repetitive pipetting. 8 or 12 identical volumes for titration plates.
- Highest possible accuracy and precision
- Easy readable digital volume display
- Volume ranges colour coded on the push button
- Volumes steplessly adjustable
- Tip ejector can be blocked to avoid mis-application when using the instrument over large series
: Each pipette is individually tested and delivered with a test certificate.
Product Family Options
8-channel
- DescrizioneVolumeIncrementoAccuratezzaImprecisione
- ME 100,5 - 10 µl0,1 µl≤ ±1,5 - ±4,0 %≤ ±1,5 - ±4,0 %1 * 1 STSFr. 621.00
- ME 505 - 501,0 µl≤ ±1,0 - ±3,0 %≤ ±0,5 - ±2,0 %1 * 1 STSFr. 621.00
- ME 30050 - 300 µl1,0 µl≤ ±0,7 - ±1,5 %≤ ±0,25 - ±0,8 %1 * 1 STSFr. 621.00
12-channel
- DescrizioneVolumeIncrementoAccuratezzaImprecisione
- ME 100,5 - 10 µl0,1 µl≤ ±1,5 - ±4,0 %≤ ±1,5 - ±4,0 %1 * 1 STSFr. 646.00
- ME 505 - 50 µl1,0 µl≤ ±3,0 (5 µl); ≤ ±1,2 (10 µl) %≤ ±0,5 - ±2,0 %1 * 1 STSFr. 646.00
- ME 30050 - 300 µl1,0 µl≤ ±0,7 - ±1,5 %≤ ±0,25 - ±0,8 %1 * 1 STSFr. 646.00