Measuring cylinders, conical, class B, borosilicate glass
Catalog # WITG4.800.001
Supplier: witeg Labortechnik
Specifications
- Capacità:1000 ml
- Divisione:20 ml
- Conf:1
Specifications
About this item
Borosilicate glass 3,3 class B.
- Excellent chemical resistance
- Good thermal resistance
- Fill volume is easy to read
Ideal for a wide range of applications. With metric graduations and spout. With a broad base for stability. Blue graduations.