ContactUsLinkComponent
Accessories for Dean-Stark, water estimators
  WITG2.892.002
undefined
Accessories for Dean-Stark, water estimators
  WITG2.892.002
  • Dimensione cono:
    29/32 NS
  • Cono femmina:
    29/32 NS
  • Descrizione:
    Measuring tube 10 ml : 0,1 ml, with stopcock, for Dean-Stark water estimator
  • Environmentally Preferable:
  • Conf:
    1