ContactUsLinkComponent
Condensers, Dimroth, double coils
Condensers, Dimroth, double coils
Catalog # WITG1.750.400
Supplier:  witeg Labortechnik
undefined
Condensers, Dimroth, double coils
Catalog # WITG1.750.400
Supplier:  witeg Labortechnik
Specifications
  • Dimensione cono:
    24/29 NS
  • Cono femmina:
    24/29 NS
  • Conf:
    1
Specifications
About this item
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With two cooling coils. Water connection with two glass olives.