Shell freezers, benchtop, FreeZone®
Supplier: LABCONCO
About this item
FreeZone® benchtop shell freezers provide a compact system for prefreezing samples for lyophilization.
- ¹/₂ hp HCFC/CFC refrigeration systems cool to −40 °C for processing samples
- LED illuminates green, when the system reaches operating temperature
- Right-side mounted power switch
- Left-side mounted retractable bath drain hose
Temperature range: − 20 °C to − 42°C
Certifications: UL certified.
1 Options Available Below
Product Details & Documents
FreeZone® benchtop shell freezers provide a compact system for prefreezing samples for lyophilization.
- ¹/₂ hp HCFC/CFC refrigeration systems cool to −40 °C for processing samples
- LED illuminates green, when the system reaches operating temperature
- Right-side mounted power switch
- Left-side mounted retractable bath drain hose
Temperature range: − 20 °C to − 42°C
Certifications: UL certified.
Specifications for Product Family
Specifications
- Catalog No:
- LBCP7949030
- LBCP7949060
- Modello:
- FreeZone®
- FreeZone®
- Codice:
- LBCP7949030
- LBCP7949060
- Display:
- LED
- LED
- Peso:
- 44 kg
- 44 kg
Specifications
Product Family Options
Product Information
- DescrizioneCapacitàTemperaturaEsterno L×P×AAvailabilityPrice
- Shell freezer, FreeZone® (EU-plug)1,2 l−20...−42 °C368×561×500 mmLog in for availability1 * 1 STSFr. 6’830.00Specifications:
More
Specifications
Cat. No.LBCP7949030ModelloFreeZone®CodiceLBCP7949030DisplayLEDPeso44 kg - Shell freezer, FreeZone® (UK-plug)1,2 l−20...−42 °C368×561×500 mmDiscontinued-Specifications:
More
Specifications
Cat. No.LBCP7949060ModelloFreeZone®CodiceLBCP7949060DisplayLEDPeso44 kg