Measuring beakers, with handle
1000 ml
Codice catalogo HECH42281066
Nome del fornitore: GLASWARENFABRIK KARL HECHT
Measuring beakers, with handle
Codice catalogo HECH42281066
Nome del fornitore: GLASWARENFABRIK KARL HECHT
Vai a
Product Details & Documents
Glass, with spout and graduation.
- Content: about 1 liters
- Outer Ø: approx. 90 mm
Ideal for general laboratory use.
Specifiche
- Capacità:1000 ml