Flasks, pear shaped, acc. to Claisen, with two necks
Catalog # WITG0.783.025
Supplier: witeg Labortechnik
Specifications
- Capacità:25 ml
- Environmentally Preferable:
- Conf:1
- Colli laterali:14/23 NS
- Collo centrale:14/23 NS
Specifications
About this item
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Center necks with socked NS 14/23, side neck with cone NS 14/23.