ContactUsLinkComponent
Elastic bandage with clips
Elastic bandage with clips
Catalog # CEDE1882
Supplier:  ORKLA CARE
undefined
Elastic bandage with clips
Catalog # CEDE1882
Supplier:  ORKLA CARE
Specifications
  • Descrizione:
    Benda elastica bianca da fermare con clip, lunghezza 4 m (tirata)
  • L×L:
    4 m × 80 mm
  • Conf:
    20
Specifications
About this item
This elastic bandage with fastening clip is intended to be used for strong fixation bandaging, for support and relief bandaging, for fixation of a wound dressing, and for aftercare of distortion, dislocations, contusions, and fractures. Made of 65% cotton and 35% polyamide.
Not intended for use directly on open wounds.