Orders LinkContactUsLinkComponent
n-Dodecil-β-D-maltoside
Irritant
n-Dodecil-β-D-maltoside
Certificates
n-Dodecil-β-D-maltoside
n-Dodecil-β-D-maltoside
2 Options Available Below
Product Details & Documents
Synonyms:  DDM, Lauryl-β-D-maltoside, Dodecyl-β-D-maltopyranoside, (2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6R)-6-(Dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, N-Dodecyl-Beta-Maltoside
  • CAS Number:69227-93-6
  • Formula:C₂₄H₄₆O₁₁
  • MDL Number:MFCD00043012
  • Molecular Weight:510.6 g/mol
  • Storage Temperature:Congelamento
Product Family Options
Product Information
  • Tipo di conf.
    Conf
    Availability
    Price
  • Bottiglia di vetro
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - SFr. 271.00
    SFr. 271.00
    1 * 1 g
       SFr. 271.00
  • Bottiglia di vetro
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - SFr. 885.00
    SFr. 885.00
    1 * 5 g
       SFr. 885.00