VWR®, dTTP solution
Catalog # E542-40UM
Supplier: VWR Chemicals
CAS Number:
Specifications
- Conf:40 µmol
- Descrizione:dTTP solution, 100 mM solution, pH 7,0
Specifications
About this item
Deoxyribonucleotides are the foundation of DNA and it takes thousands of these nucleotides to make up DNA.
- Building blocks for the amplification of DNA
- High purity grade
DNA consists of a sugar molecule (deoxyribose), a nitrogenous base (purine in adenine and guanine, pyrimidine in thymine or cytosine) and a phosphate molecule.