Order Entry
Switzerland
ContactUsLinkComponent
Amido nero 10B ~50% (dye content), Sigma-Aldrich®
Irritant
Amido nero 10B ~50% (dye content), Sigma-Aldrich®
Catalog # N9002-100G
CAS Number:  
undefined
Irritant
Amido nero 10B ~50% (dye content), Sigma-Aldrich®
Catalog # N9002-100G
CAS Number:  

Specifications

  • Tipo di conf.:
    Bottiglia di vetro
  • Conf:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Appearance (Form):
    Powder
  • Solubility (Colour):
    Dark Blue
  • Nitrogen:
    5.5 - 8.2%
  • Lambda Max, In H₂O:
    617 - 620 nm
  • Dye Content:
    40 - 60%, Approx 50%

Specification Test Results

About this item

 :
 :616.49 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:213-903-1
 :GHS07
Storage Temperature:Room Temperature
CAS Number:1064-48-8
Formula:C₂₂H₁₄N₆Na₂O₉S₂
MDL Number:MFCD00004017