Verde Fast
Glass bottle for solids, 10 g
Catalog # NADIHS-606-10G
Marketsource
Supplier: NATIONAL DIAGNOSTICS
Jump to:
Product Details & Documents
Synonyms: CI 42590, Ethyl Green, Methyl green zinc chloride double salt, Methyl Green
- CAS Number:7114-03-6
- Formula:C₂₇H₃₅BrClN₃·xZnCl₂
- MDL Number:MFCD00151094
- Molecular Weight:653.24 g/mol
- Storage Temperature:Ambiente
- EINECS:230-415-4
Specifications
Specifications
- Tipo di conf.:Glass bottle for solids
- Conf:10 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



