Wound spreaders
Wound spreader, Scalp-Contour, self-retaining, blunt ends, stainless steel 18/8, 135 mm, 55 mm
Catalog # MART15-727-14
Supplier: KLS Martin Group
Jump to:
Product Details & Documents
Stainless steel 18/8.
Specifications
- Larghezza di serraggio:55 mm
- Descrizione:Wound spreader, Scalp-Contour, self-retaining, blunt ends, stainless steel 18/8
- Lunghezza:135 mm