Capillary columns, Elite-35 for autosystem
Nome del fornitore: PerkinElmer
Vai a
Informazioni su questo articolo
Elite-35 Columns are dimethylpolysiloxane (35% Diphenyl) silica fused columns; the small percentage of phenyl groups imparts selectivity towards aromatic components.
4 varianti
Product Details & Documents
Elite-35 Columns are dimethylpolysiloxane (35% Diphenyl) silica fused columns; the small percentage of phenyl groups imparts selectivity towards aromatic components.
Product Family Options
Informazioni sul prodotto
- Formato della colonnaFaseSpessore dello stratoØ int.LunghezzaDisponibilitàPrice