Thermal printer
CONSAP414
: CONSORT
- Descrizione:Stampante seriale, matrice termica a 9×320 punti
- Conf:1
This thermal printer can deliver both serial and parallel input when using RS232 and Centronix connectors respectively. Direct connection is available to connect all meters with digital output.
- Prints 40 columns in normal and 80 columns in condensed condition
- Roll length: ±28 m
: Supplied with manual, roll of thermal paper, mains adaptor (230 VAC) and RS232 cable.