Beakers for storing micro instruments
75 mm, 75 mm
Codice catalogo MART55-348-00
Nome del fornitore: KLS Martin Group
Beakers for storing micro instruments
Codice catalogo MART55-348-00
Nome del fornitore: KLS Martin Group
Vai a
Product Details & Documents
Stainless steel.
- Good chemical resistance
- Excellent resistance to heat
- Smooth surfaces for fast, easy cleaning
For storing micro instruments.
Specifiche
- Ø est:75 mm
- Altezza:75 mm