Orders LinkContactUsLinkComponent
Neutralisation agent, Helimatic® Neutralizer C
Neutralisation agent, Helimatic® Neutralizer C
Helimatic® Neutralizer C (based on citric acid), 5 l
Codice catalogo BRAU18732
Nome del fornitore:   B. Braun
Neutralisation agent, Helimatic® Neutralizer C
Codice catalogo BRAU18732
Nome del fornitore:  B. Braun
Neutralisation agent, Helimatic® Neutralizer C
Neutralisation agent, Helimatic® Neutralizer C
Helimatic® Neutralizer C (based on citric acid), 5 l
Price
139.00 SFr.
Log in per disponibilità
1 * 5 kg
Product Details & Documents
Helimatic® Neutralizer C is a liquid neutraliser containing citric acid which is used for the neutralisation of alkaline resides during machine processing of instruments. It has a low working concentration from 0,05% - 0,3%.
  • Automatic reprocessing of resuable invasive and non invasive medical devices
  • Free of phosphates and surfactants
  • Based on citric acid (pharmaceutical grade)
Documents
Specifiche
  • Capacità:
    5 l
  • Descrizione:
    Helimatic® Neutralizer C (based on citric acid)