Face screen without carrier, S591
Face screen without carrier, Chemical resistant, Acetato
Codice catalogo CENUS591
Nome del fornitore: CENTURION SAFETY PRODUCTS
Face screen without carrier, S591
Codice catalogo CENUS591
Nome del fornitore: CENTURION SAFETY PRODUCTS
Vai a
Product Details & Documents
Long acetate face screen, to be combined with the carrier S54 for helmet attachment.
- Resistant against organic and inorganic chemicals
- Ideal for metal cutting applications
Length: 210 mm
Certifications: EN 166
Specifiche
- Tipo:Chemical resistant
- Descrizione:Face screen without carrier
- Materiale:Acetato